Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr.10: 91923769 - 92030998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003972.2 NP_003963.1 11 - 12 1361 107
XM_011540326.1 XP_011538628.1 11 - 12 1571 107
XM_011540327.2 XP_011538629.1 8 - 9 1126 107
XM_011540328.2 XP_011538630.1 8 - 9 954 107
XM_011540329.2 XP_011538631.1 11 - 12 1571 107
XM_017016877.1 XP_016872366.1 10 - 11 1423 107
XM_017016878.1 XP_016872367.1 10 - 11 1253 107
XM_017016879.1 XP_016872368.1 10 - 11 1377 107
XM_017016880.1 XP_016872369.1 10 - 11 1381 107
XM_017016881.1 XP_016872370.1 9 - 10 1230 107
GenBank mRNA AF038362.1 - 11 - 12 1380 107
AJ001017.2 - 11 - 12 1275 107
BC030273.1 - 5 - 6 608 107
BC112201.1 - 11 - 12 1345 107

View Full Product Details