The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080527.1 | NP_001073996.1 | 29 - 30 | 4023 | 55 |
XM_006712539.3 | XP_006712602.1 | 30 - 31 | 4214 | 55 | |
XM_011511218.2 | XP_011509520.1 | 30 - 31 | 4223 | 55 | |
XM_017004169.1 | XP_016859658.1 | 30 - 31 | 4223 | 55 | |
GenBank mRNA | AB290182.1 | - | 5 - 6 | 877 | 55 |
AK074183.1 | - | 4 - 5 | 955 | 55 | |
CR627350.1 | - | - | 823 | 55 |