The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080527.1 | NP_001073996.1 | 31 - 32 | 4309 | 78 |
XM_006712539.3 | XP_006712602.1 | 32 - 33 | 4500 | 78 | |
XM_011511218.2 | XP_011509520.1 | 32 - 33 | 4509 | 78 | |
GenBank mRNA | AB290182.1 | - | 7 - 8 | 1163 | 78 |
AK074183.1 | - | 6 - 7 | 1241 | 78 |