Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPOCD1
Entrez Gene ID
90853
Gene Name
SPOC domain containing 1
Gene Aliases
PPP1R146
Chromosome Location
Chr.1: 31790422 - 31816051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281987.1 NP_001268916.1 10 - 11 2405 66
NM_001281988.1 NP_001268917.1 9 - 10 983 66
NM_144569.5 NP_653170.3 10 - 11 2405 66
XM_011542413.2 XP_011540715.1 10 - 11 974 66
XM_011542414.2 XP_011540716.1 9 - 10 1169 66
XM_017002780.1 XP_016858269.1 8 - 9 2272 66
XM_017002782.1 XP_016858271.1 9 - 10 1543 66
GenBank mRNA AK091016.1 - 9 - 10 954 66
AK131447.1 - 10 - 11 2334 66
BC114430.1 - 10 - 11 2331 66

View Full Product Details