The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174922.4 | NP_777582.4 | 6 - 7 | 762 | 63 |
XM_006716527.2 | XP_006716590.1 | 6 - 7 | 752 | 63 | |
XM_011516907.2 | XP_011515209.1 | 6 - 7 | 1475 | 63 | |
XM_011516909.1 | XP_011515211.1 | 6 - 7 | 762 | 63 | |
XM_011516910.1 | XP_011515212.1 | 7 - 8 | 785 | 63 | |
XM_017013174.1 | XP_016868663.1 | 6 - 7 | 800 | 63 | |
GenBank mRNA | AK092773.1 | - | 6 - 7 | 725 | 63 |
BC085013.1 | - | 1 - 2 | 72 | 63 | |
BC101659.1 | - | 6 - 7 | 702 | 63 | |
BC143326.1 | - | 5 - 6 | 529 | 63 | |
BU845476.1 | - | 1 - 2 | 70 | 63 |