Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2F7
Entrez Gene ID
144455
Gene Name
E2F transcription factor 7
Gene Aliases
-
Chromosome Location
Chr.12: 77021246 - 77065580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203394.2 NP_976328.2 10 - 11 2119 87
XM_011537966.2 XP_011536268.1 9 - 10 1984 87
XM_011537969.2 XP_011536271.1 9 - 10 1682 87
GenBank mRNA AK096316.1 - 1 - 2 376 87
AK097677.1 - 8 - 9 1710 87
BC016658.1 - 1 - 2 103 87
BC136366.1 - 10 - 11 1958 87
BC136367.1 - 10 - 11 1958 87

View Full Product Details