The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006959.2 | NP_008890.2 | 1 - 2 | 246 | 59 |
GenBank mRNA | AK098183.1 | - | 1 - 2 | 228 | 59 |