Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
Chromosome Location
Chr.12: 121402289 - 121582268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005366.1 NP_001005366.1 6 - 7 1000 71
NM_032590.4 NP_115979.3 6 - 7 790 71
XM_005253955.3 XP_005254012.1 6 - 7 1000 71
XM_005253956.3 XP_005254013.1 6 - 7 597 71
XM_011538867.2 XP_011537169.1 6 - 7 2043 71
XM_011538868.2 XP_011537170.1 6 - 7 2039 71
XM_011538869.2 XP_011537171.1 6 - 7 2041 71
GenBank mRNA AJ459424.1 - 6 - 7 762 71
AK127328.1 - 6 - 7 1000 71
AL133572.1 - 2 - 3 303 71
BC115379.1 - 5 - 6 650 71
BC115380.1 - 6 - 7 729 71
BX648928.1 - 6 - 7 792 71

View Full Product Details