The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308152.1 | NP_001295081.1 | 5 - 6 | 733 | 147 |
NM_001308153.1 | NP_001295082.1 | 6 - 7 | 822 | 147 | |
NM_014629.3 | NP_055444.2 | 5 - 6 | 730 | 147 | |
XM_005266041.3 | XP_005266098.1 | 5 - 6 | 672 | 147 | |
XM_011534767.2 | XP_011533069.1 | 4 - 5 | 592 | 147 | |
XM_011534768.1 | XP_011533070.1 | 4 - 5 | 595 | 147 | |
XM_011534770.2 | XP_011533072.1 | 4 - 5 | 595 | 147 | |
XM_017014003.1 | XP_016869492.1 | 5 - 6 | 635 | 147 | |
GenBank mRNA | AB002292.2 | - | 5 - 6 | 729 | 147 |
AK309799.1 | - | 3 - 4 | 461 | 147 | |
AL137508.1 | - | 2 - 3 | 334 | 146 | |
BC112926.1 | - | 5 - 6 | 721 | 147 | |
CR749570.1 | - | 6 - 7 | 783 | 147 |