Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
Chromosome Location
Chr.12: 1791957 - 1922606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172364.4 NP_758952.4 35 - 36 3341 78
XM_011521041.2 XP_011519343.1 34 - 35 7180 78
GenBank mRNA AF516695.1 - 35 - 36 3473 78
AK301234.1 - 8 - 9 582 78
AL137658.1 - 12 - 13 880 78
BC048288.1 - 7 - 8 447 78
BC150186.1 - 34 - 35 3264 78
BI908421.1 - 8 - 9 549 78
BX537436.1 - 33 - 34 3941 78
BX537437.1 - 35 - 36 78

View Full Product Details