Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STRADA
Entrez Gene ID
92335
Gene Name
STE20-related kinase adaptor alpha
Gene Aliases
LYK5, NY-BR-96, PMSE, STRAD, Stlk
Chromosome Location
Chr.17: 63702832 - 63741970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003786.2 NP_001003786.1 6 - 7 739 60
NM_001003787.2 NP_001003787.1 8 - 9 850 60
NM_001003788.2 NP_001003788.1 7 - 8 821 60
NM_001165969.1 NP_001159441.1 6 - 7 763 60
NM_001165970.1 NP_001159442.1 6 - 7 718 60
NM_153335.5 NP_699166.2 6 - 7 739 60
XM_005257797.2 XP_005257854.1 8 - 9 701 60
XM_005257798.2 XP_005257855.1 6 - 7 663 60
XM_005257799.2 XP_005257856.1 7 - 8 672 60
XM_005257800.2 XP_005257857.1 8 - 9 725 60
XM_005257801.4 XP_005257858.1 8 - 9 725 60
XM_011525466.2 XP_011523768.1 8 - 9 725 60
XM_011525467.2 XP_011523769.1 8 - 9 752 60
XM_017025312.1 XP_016880801.1 6 - 7 614 60
XM_017025313.1 XP_016880802.1 7 - 8 696 60
XM_017025314.1 XP_016880803.1 6 - 7 663 60
GenBank mRNA AF308302.1 - 8 - 9 689 60
AK074771.1 - 6 - 7 605 60
AK075005.1 - 6 - 7 568 60
AK293160.1 - 6 - 7 626 60
AK301331.1 - 6 - 7 590 60
AL832407.1 - 7 - 8 634 60
AY290821.2 - 6 - 7 739 60
BC043641.1 - 7 - 8 612 60

View Full Product Details