The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040424.2 | NP_001035514.1 | 19 - 20 | 2613 | 75 |
NM_001282934.1 | NP_001269863.1 | 20 - 21 | 2673 | 75 | |
NM_022115.4 | NP_071398.3 | 26 - 27 | 3600 | 75 | |
NR_104257.1 | 20 - 21 | 2621 | 75 | ||
NR_104258.1 | 19 - 20 | 2501 | 75 | ||
NR_104260.1 | 19 - 20 | 2468 | 75 | ||
NR_135464.1 | 19 - 20 | 2609 | 75 | ||
XM_006724039.2 | XP_006724102.1 | 19 - 20 | 2455 | 75 | |
XM_006724040.2 | XP_006724103.1 | 18 - 19 | 2395 | 75 | |
XM_011529674.1 | XP_011527976.1 | 19 - 20 | 2485 | 75 | |
XM_011529675.1 | XP_011527977.1 | 20 - 21 | 2529 | 75 | |
XM_011529676.2 | XP_011527978.1 | 20 - 21 | 4938 | 75 | |
XM_011529677.2 | XP_011527979.1 | 20 - 21 | 2614 | 75 | |
XM_011529678.2 | XP_011527980.1 | 20 - 21 | 2658 | 75 | |
XM_011529679.2 | XP_011527981.1 | 20 - 21 | 2991 | 75 | |
XM_011529681.2 | XP_011527983.1 | 20 - 21 | 3895 | 75 | |
XM_011529683.1 | XP_011527985.1 | 25 - 26 | 3316 | 75 | |
XM_017028425.1 | XP_016883914.1 | 19 - 20 | 2468 | 75 | |
XM_017028426.1 | XP_016883915.1 | 19 - 20 | 4877 | 75 | |
GenBank mRNA | AB051812.1 | - | 19 - 20 | 2604 | 75 |
AB051813.1 | - | 20 - 21 | 2664 | 75 | |
AB051814.1 | - | 20 - 21 | 2612 | 75 | |
AB051815.1 | - | 19 - 20 | 2492 | 75 | |
AY063456.1 | - | 1 - 2 | 75 | ||
AY078498.2 | - | 26 - 27 | 3600 | 75 | |
BC067102.1 | - | 19 - 20 | 2468 | 75 |