The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175571.3 | NP_783161.1 | 2 - 3 | 1213 | 79 |
XM_005249950.4 | XP_005250007.1 | 2 - 3 | 893 | 79 | |
XM_005249951.3 | XP_005250008.1 | 2 - 3 | 1213 | 79 | |
GenBank mRNA | AJ633686.1 | - | 1 - 2 | 639 | 79 |
AK098320.1 | - | 1 - 2 | 398 | 79 | |
AK314426.1 | - | 2 - 3 | 1048 | 79 | |
AL834361.1 | - | 2 - 3 | 807 | 79 | |
BC107037.2 | - | 2 - 3 | 800 | 79 |