The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033553.2 | NP_001028725.1 | 10 - 11 | 2731 | 63 |
NM_001033555.2 | NP_001028727.1 | 8 - 9 | 2640 | 63 | |
NM_001243439.1 | NP_001230368.1 | 10 - 11 | 2790 | 63 | |
XM_005256860.2 | XP_005256917.1 | 7 - 8 | 1060 | 63 | |
XM_017025317.1 | XP_016880806.1 | 9 - 10 | 2707 | 63 | |
XM_017025318.1 | XP_016880807.1 | 9 - 10 | 2707 | 63 | |
XM_017025319.1 | XP_016880808.1 | 10 - 11 | 2849 | 63 | |
XM_017025320.1 | XP_016880809.1 | 10 - 11 | 2825 | 63 | |
XM_017025321.1 | XP_016880810.1 | 8 - 9 | 2664 | 63 | |
GenBank mRNA | AK295093.1 | - | 10 - 11 | 2763 | 63 |
AK295811.1 | - | 7 - 8 | 2267 | 63 | |
AK298601.1 | - | 7 - 8 | 1472 | 63 | |
AK301318.1 | - | 7 - 8 | 982 | 63 | |
AK304186.1 | - | 8 - 9 | 2562 | 63 | |
AY816327.1 | - | 8 - 9 | 2545 | 63 | |
AY816329.1 | - | 10 - 11 | 2731 | 63 | |
BC021123.1 | - | 8 - 9 | 2538 | 63 | |
CN267630.1 | - | 2 - 3 | 106 | 63 |