The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172435.1 | NP_001165906.1 | 20 - 21 | 2430 | 104 |
NM_012233.2 | NP_036365.1 | 20 - 21 | 2430 | 104 | |
XM_011510823.2 | XP_011509125.1 | 20 - 21 | 2430 | 104 | |
XM_011510825.2 | XP_011509127.1 | 20 - 21 | 2430 | 104 | |
GenBank mRNA | AB449877.1 | - | 20 - 21 | 2387 | 104 |
AK022494.1 | - | 20 - 21 | 2402 | 104 | |
AL096752.1 | - | 1 - 2 | 1156 | 104 | |
BC022977.1 | - | 7 - 8 | 1074 | 104 | |
BC036732.1 | - | 3 - 4 | 499 | 104 | |
BC146809.1 | - | 22 - 23 | 2667 | 104 | |
D31886.1 | - | 20 - 21 | 2389 | 104 |