The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015155.2 | NP_055970.1 | 17 - 18 | 2246 | 57 |
XM_017015988.1 | XP_016871477.1 | 16 - 17 | 4293 | 57 | |
XM_017015994.1 | XP_016871483.1 | 16 - 17 | 1932 | 57 | |
GenBank mRNA | BC131630.1 | - | 16 - 17 | 1963 | 57 |
BC152443.1 | - | 16 - 17 | 1970 | 57 | |
CR749653.1 | - | 3 - 4 | 4420 | 57 | |
D86971.2 | - | 16 - 17 | 1970 | 57 |