The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319085.1 | NP_001306014.1 | 1 - 2 | 118 | 60 |
NM_001319087.1 | NP_001306016.1 | 1 - 2 | 118 | 60 | |
NM_001319094.1 | NP_001306023.1 | 1 - 2 | 118 | 60 | |
NM_001319096.1 | NP_001306025.1 | 1 - 2 | 118 | 60 | |
NM_212481.2 | NP_997646.1 | 1 - 2 | 118 | 60 | |
XM_017003184.1 | XP_016858673.1 | 1 - 2 | 84 | 60 | |
GenBank mRNA | AK300173.1 | - | 1 - 2 | 85 | 60 |
AK304566.1 | - | 1 - 2 | 85 | 60 | |
AK307768.1 | - | 1 - 2 | 88 | 60 | |
BC067301.1 | - | 1 - 2 | 65 | 60 | |
BM556343.1 | - | 1 - 2 | 63 | 60 | |
BQ073595.1 | - | - | 123 | 60 | |
BX396215.2 | - | 1 - 2 | 68 | 60 | |
BX648918.1 | - | 1 - 2 | 86 | 60 | |
CR981009.1 | - | 1 - 2 | 73 | 60 | |
HY002073.1 | - | 1 - 2 | 118 | 60 | |
M62324.1 | - | 1 - 2 | 70 | 60 |