The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152513.3 | NP_689726.3 | 24 - 25 | 3112 | 79 |
XM_011529940.1 | XP_011528242.1 | 23 - 24 | 3016 | 79 | |
XM_011529941.1 | XP_011528243.1 | 24 - 25 | 3121 | 79 | |
XM_017028633.1 | XP_016884122.1 | 24 - 25 | 3121 | 79 | |
GenBank mRNA | AK092934.1 | - | 10 - 11 | 1481 | 79 |
AK093341.1 | - | 4 - 5 | 617 | 79 | |
AK094461.1 | - | 6 - 7 | 898 | 79 | |
AK097834.1 | - | 4 - 5 | 663 | 79 | |
AK128158.1 | - | 8 - 9 | 5836 | 79 | |
AK301391.1 | - | 1 - 2 | 431 | 79 | |
AK301435.1 | - | 9 - 10 | 1318 | 79 | |
AK307341.1 | - | 1 - 2 | 481 | 79 | |
AY952376.1 | - | 9 - 10 | 1824 | 79 | |
AY952377.1 | - | 8 - 9 | 1719 | 79 | |
BC029582.1 | - | 5 - 6 | 861 | 79 | |
BC032248.1 | - | 1 - 2 | 446 | 79 | |
BC070111.1 | - | 20 - 21 | 2762 | 79 |