The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099437.1 | NP_001092907.1 | 4 - 5 | 636 | 66 |
NM_001099438.1 | NP_001092908.1 | 4 - 5 | 702 | 66 | |
NM_194325.2 | NP_919306.2 | 4 - 5 | 633 | 66 | |
XM_011527443.2 | XP_011525745.1 | 4 - 5 | 752 | 66 | |
XM_011527444.1 | XP_011525746.1 | 4 - 5 | 341 | 66 | |
XM_017027423.1 | XP_016882912.1 | 3 - 4 | 961 | 66 | |
XM_017027424.1 | XP_016882913.1 | 4 - 5 | 387 | 66 | |
XM_017027425.1 | XP_016882914.1 | 4 - 5 | 692 | 66 | |
XM_017027426.1 | XP_016882915.1 | 4 - 5 | 1148 | 66 | |
GenBank mRNA | AK295515.1 | - | 4 - 5 | 492 | 66 |
BC142996.1 | - | 4 - 5 | 621 | 66 |