Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF33
Entrez Gene ID
100271715
Gene Name
Rho guanine nucleotide exchange factor 33
Gene Aliases
-
Chromosome Location
Chr.2: 38919363 - 38975449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AW468668.1 - 2 - 3 268 69

View Full Product Details