The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207307.2 | NP_997190.1 | 1 - 2 | 214 | 71 |
GenBank mRNA | AK127968.1 | - | 1 - 2 | 199 | 71 |
BC066975.1 | - | 1 - 2 | 214 | 71 |