The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301339.1 | NP_001288268.1 | 2 - 3 | 125 | |
GenBank mRNA | BM128374.1 | - | 2 - 3 | 360 | 125 |