Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAGE3
Entrez Gene ID
139793
Gene Name
PAGE family member 3
Gene Aliases
CT16.6, GAGED1, PAGE-3
Chromosome Location
Chr.X: 55257833 - 55264916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017931.2 NP_001017931.2 3 - 4 501 108
NM_001171252.1 NP_001164723.1 3 - 4 515 108
NM_001303613.1 NP_001290542.1 3 - 4 303 108
XM_017029282.1 XP_016884771.1 3 - 4 304 108
GenBank mRNA BC140001.1 - 3 - 4 374 108
BC140741.1 - 3 - 4 387 108
BC144449.1 - 3 - 4 400 108
HY049011.1 - 3 - 4 303 108

View Full Product Details