The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001740.4 | NP_001731.2 | 1 - 2 | 177 | 107 |
NM_007088.3 | NP_009019.1 | 1 - 2 | 177 | 107 | |
NR_027910.2 | 1 - 2 | 177 | 107 | ||
GenBank mRNA | AK222495.1 | - | 1 - 2 | 167 | 107 |
AK222648.1 | - | 1 - 2 | 167 | 107 | |
AK291096.1 | - | 1 - 2 | 167 | 107 | |
BC015484.2 | - | 1 - 2 | 152 | 107 | |
DA060805.1 | - | 1 - 2 | 177 | 107 | |
X56667.1 | - | 1 - 2 | 140 | 107 |