Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B4GALNT4
Entrez Gene ID
338707
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.11: 369773 - 382117 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178537.4 NP_848632.2 3 - 4 362 76
XM_017017654.1 XP_016873143.1 3 - 4 1674 76
GenBank mRNA AB089939.1 - 3 - 4 353 76
BC131528.1 - 3 - 4 349 76
BF058197.1 - 3 - 4 362 76

View Full Product Details