The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170710.4 | NP_733828.2 | 28 - 29 | 3874 | 105 |
NM_181265.3 | NP_851782.3 | 26 - 27 | 3706 | 105 | |
XM_005262725.3 | XP_005262782.1 | 26 - 27 | 3373 | 105 | |
XM_011531582.2 | XP_011529884.1 | 27 - 28 | 3715 | 105 | |
XM_017007686.1 | XP_016863175.1 | 27 - 28 | 3703 | 105 | |
XM_017007687.1 | XP_016863176.1 | 28 - 29 | 3788 | 105 | |
XM_017007688.1 | XP_016863177.1 | 28 - 29 | 3705 | 105 | |
XM_017007689.1 | XP_016863178.1 | 26 - 27 | 3681 | 105 | |
XM_017007690.1 | XP_016863179.1 | 27 - 28 | 3767 | 105 | |
XM_017007691.1 | XP_016863180.1 | 26 - 27 | 3537 | 105 | |
XM_017007692.1 | XP_016863181.1 | 27 - 28 | 3623 | 105 | |
XM_017007693.1 | XP_016863182.1 | 25 - 26 | 3516 | 105 | |
XM_017007694.1 | XP_016863183.1 | 26 - 27 | 3500 | 105 | |
XM_017007695.1 | XP_016863184.1 | 26 - 27 | 3551 | 105 | |
XM_017007696.1 | XP_016863185.1 | 25 - 26 | 3530 | 105 | |
GenBank mRNA | AF492460.1 | - | 28 - 29 | 3727 | 105 |
AK130128.1 | - | 12 - 13 | 1336 | 105 | |
BX648508.1 | - | 12 - 13 | 1274 | 105 |