Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52314168-52314727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT83
Entrez Gene ID
3889
Gene Name
keratin 83
Gene Aliases
HB3, Hb-3, KRTHB3
UniGene
Hs.720768
Gene Chromosome Location
Chr.12: 52314301-52321398 on Build GRCh38
Transcripts
NM_002282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419048 c.1477C>T p.H493Y Chr.12: 52314636-52314636 on Build GRCh38
3987010 c.1429G>A p.G477S Chr.12: 52314684-52314684 on Build GRCh38
7374426 c.1457G>C p.G486A Chr.12: 52314656-52314656 on Build GRCh38
6454856 c.1464C>T p.C488C Chr.12: 52314649-52314649 on Build GRCh38
3462209 c.1424C>T p.P475L Chr.12: 52314689-52314689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944733057 Chr.12: 52314196-52314196 on Build GRCh38
rs955581900 Chr.12: 52314529-52314529 on Build GRCh38
rs970112283 Chr.12: 52314241-52314241 on Build GRCh38
rs572039958 Chr.12: 52314294-52314294 on Build GRCh38
rs563097150 Chr.12: 52314402-52314402 on Build GRCh38
rs535186700 Chr.12: 52314608-52314608 on Build GRCh38
rs546378595 Chr.12: 52314353-52314353 on Build GRCh38
rs1036430232 Chr.12: 52314451-52314451 on Build GRCh38
rs190585205 Chr.12: 52314629-52314629 on Build GRCh38
rs2857671 Chr.12: 52314636-52314636 on Build GRCh38
rs777701732 Chr.12: 52314673-52314673 on Build GRCh38
rs910318322 Chr.12: 52314347-52314347 on Build GRCh38
rs748930934 Chr.12: 52314704-52314704 on Build GRCh38
rs886049620 Chr.12: 52314406-52314406 on Build GRCh38
rs779718237 Chr.12: 52314702-52314702 on Build GRCh38
rs367824594 Chr.12: 52314695-52314695 on Build GRCh38
rs989100936 Chr.12: 52314509-52314509 on Build GRCh38
rs959802327 Chr.12: 52314308-52314308 on Build GRCh38
rs758618491 Chr.12: 52314665-52314665 on Build GRCh38
rs771547026 Chr.12: 52314524-52314524 on Build GRCh38
rs2852456 Chr.12: 52314457-52314457 on Build GRCh38
rs765607233 Chr.12: 52314653-52314653 on Build GRCh38
rs752120043 Chr.12: 52314681-52314681 on Build GRCh38
rs2852455 Chr.12: 52314268-52314268 on Build GRCh38
rs755878013 Chr.12: 52314352-52314352 on Build GRCh38
rs956056629 Chr.12: 52314496-52314496 on Build GRCh38
rs964824702 Chr.12: 52314588-52314588 on Build GRCh38
rs888146161 Chr.12: 52314272-52314272 on Build GRCh38
rs2857672 Chr.12: 52314493-52314493 on Build GRCh38
rs531678283 Chr.12: 52314404-52314404 on Build GRCh38
rs563887012 Chr.12: 52314648-52314648 on Build GRCh38
rs114956210 Chr.12: 52314341-52314341 on Build GRCh38
rs895646596 Chr.12: 52314188-52314188 on Build GRCh38
rs571944353 Chr.12: 52314635-52314635 on Build GRCh38
rs527729235 Chr.12: 52314481-52314481 on Build GRCh38
rs186532103 Chr.12: 52314587-52314587 on Build GRCh38
rs11285858 Chr.12: 52314307-52314307 on Build GRCh38
rs767352531 Chr.12: 52314642-52314642 on Build GRCh38
rs1036547068 Chr.12: 52314225-52314225 on Build GRCh38
rs539354383 Chr.12: 52314553-52314553 on Build GRCh38
rs768266558 Chr.12: 52314708-52314708 on Build GRCh38
rs182482854 Chr.12: 52314269-52314269 on Build GRCh38
rs777492822 Chr.12: 52314363-52314363 on Build GRCh38
rs555952638 Chr.12: 52314190-52314190 on Build GRCh38
rs929248882 Chr.12: 52314401-52314401 on Build GRCh38
rs1049664438 Chr.12: 52314460-52314460 on Build GRCh38
rs577585131 Chr.12: 52314649-52314649 on Build GRCh38
rs762496002 Chr.12: 52314632-52314632 on Build GRCh38
rs1000243240 Chr.12: 52314479-52314479 on Build GRCh38
rs200232339 Chr.12: 52314690-52314690 on Build GRCh38
rs879485197 Chr.12: 52314267-52314268 on Build GRCh38
rs548698510 Chr.12: 52314456-52314456 on Build GRCh38
rs1013979954 Chr.12: 52314555-52314555 on Build GRCh38
rs377671303 Chr.12: 52314424-52314425 on Build GRCh38
rs560812760 Chr.12: 52314316-52314316 on Build GRCh38
rs147359482 Chr.12: 52314701-52314701 on Build GRCh38
rs749668667 Chr.12: 52314355-52314355 on Build GRCh38
rs374145143 Chr.12: 52314503-52314503 on Build GRCh38
rs386763038 Chr.12: 52314267-52314268 on Build GRCh38
rs749257057 Chr.12: 52314381-52314381 on Build GRCh38
rs761601312 Chr.12: 52314637-52314637 on Build GRCh38
rs570369664 Chr.12: 52314514-52314514 on Build GRCh38
rs2852454 Chr.12: 52314267-52314267 on Build GRCh38
rs549538781 Chr.12: 52314574-52314574 on Build GRCh38
rs867309581 Chr.12: 52314668-52314668 on Build GRCh38
rs371464628 Chr.12: 52314586-52314586 on Build GRCh38
rs979184347 Chr.12: 52314247-52314247 on Build GRCh38
rs944802096 Chr.12: 52314434-52314434 on Build GRCh38
rs199721779 Chr.12: 52314310-52314310 on Build GRCh38
rs778405583 Chr.12: 52314612-52314612 on Build GRCh38
rs2857673 Chr.12: 52314189-52314189 on Build GRCh38
rs752938418 Chr.12: 52314656-52314656 on Build GRCh38
rs757638030 Chr.12: 52314688-52314688 on Build GRCh38
rs771384854 Chr.12: 52314581-52314581 on Build GRCh38
rs1019987441 Chr.12: 52314570-52314570 on Build GRCh38
rs201427021 Chr.12: 52314297-52314297 on Build GRCh38
rs986150676 Chr.12: 52314423-52314423 on Build GRCh38
rs895676012 Chr.12: 52314410-52314410 on Build GRCh38
rs555026767 Chr.12: 52314285-52314285 on Build GRCh38
rs1014028730 Chr.12: 52314411-52314411 on Build GRCh38

View Full Product Details