Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAN2C1
Entrez Gene ID
4123
Gene Name
mannosidase alpha class 2C member 1
Gene Aliases
MAN6A8, MANA, MANA1
Chromosome Location
Chr.15: 75355792 - 75368680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256494.1 NP_001243423.1 6 - 7 834 65
NM_001256495.1 NP_001243424.1 6 - 7 834 65
NM_006715.3 NP_006706.2 6 - 7 834 65
XM_005254384.2 XP_005254441.1 6 - 7 887 65
XM_017022186.1 XP_016877675.1 5 - 6 1182 65
XM_017022187.1 XP_016877676.1 5 - 6 1182 65
GenBank mRNA AF044414.2 - 6 - 7 846 65
AK311065.1 - 5 - 6 686 65
AL136876.1 - 6 - 7 795 65
BC050550.1 - 6 - 7 801 65
BC080191.1 - 6 - 7 799 65

View Full Product Details