The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184998.1 | NP_001171927.1 | 16 - 17 | 3467 | 101 |
NM_001184999.1 | NP_001171928.1 | 16 - 17 | 3458 | 101 | |
NM_014647.3 | NP_055462.2 | 16 - 17 | 3467 | 101 | |
XM_005255704.1 | XP_005255761.1 | 16 - 17 | 3463 | 101 | |
XM_005255706.1 | XP_005255763.1 | 16 - 17 | 3457 | 101 | |
XM_005255707.1 | XP_005255764.1 | 16 - 17 | 3433 | 101 | |
XM_017023900.1 | XP_016879389.1 | 16 - 17 | 3351 | 101 | |
XM_017023901.1 | XP_016879390.1 | 16 - 17 | 3348 | 101 | |
XM_017023902.1 | XP_016879391.1 | 15 - 16 | 3301 | 101 | |
XM_017023903.1 | XP_016879392.1 | 15 - 16 | 3034 | 101 | |
XM_017023904.1 | XP_016879393.1 | 15 - 16 | 3031 | 101 | |
XM_017023905.1 | XP_016879394.1 | 16 - 17 | 2888 | 101 | |
XM_017023906.1 | XP_016879395.1 | 16 - 17 | 2829 | 101 | |
GenBank mRNA | AB007890.3 | - | 14 - 15 | 2553 | 101 |
BC137165.1 | - | 16 - 17 | 3409 | 101 | |
BC137170.1 | - | 16 - 17 | 3409 | 101 | |
BC144514.1 | - | 16 - 17 | 3409 | 101 | |
BC144515.1 | - | 16 - 17 | 3400 | 101 | |
CN310535.1 | - | 3 - 4 | 546 | 101 |