The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080387.4 | NP_525126.2 | 3 - 4 | 429 | 85 |
XM_011520632.2 | XP_011518934.1 | 4 - 5 | 398 | 85 | |
XM_011520633.2 | XP_011518935.1 | 3 - 4 | 424 | 85 | |
GenBank mRNA | AF411850.1 | - | 3 - 4 | 299 | 85 |
AK314071.1 | - | 3 - 4 | 416 | 85 | |
AY115592.1 | - | 3 - 4 | 420 | 85 | |
AY297446.1 | - | 3 - 4 | 428 | 85 | |
AY486482.1 | - | 3 - 4 | 253 | 85 | |
AY486483.1 | - | 3 - 4 | 253 | 85 | |
BC032313.1 | - | 3 - 4 | 429 | 85 | |
BI909298.1 | - | 3 - 4 | 378 | 85 |