Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56450528-56451106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASS4
Entrez Gene ID
57091
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
C20orf32, CAS4, HEFL, HEPL
UniGene
Hs.473144
Gene Chromosome Location
Chr.20: 56411550-56460240 on Build GRCh38
Transcripts
NM_001164114.1 NM_001164115.1 NM_001164116.1 NM_020356.3 XM_006723831.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620684 c.591A>G p.I197M Chr.20: 56450628-56450628 on Build GRCh38
4581902 c.626C>G p.P209R Chr.20: 56450663-56450663 on Build GRCh38
6160480 c.597C>A p.A199A Chr.20: 56450634-56450634 on Build GRCh38
1751510 c.568G>A p.E190K Chr.20: 56450605-56450605 on Build GRCh38
359711 c.630C>A p.D210E Chr.20: 56450667-56450667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201734849 Chr.20: 56450579-56450579 on Build GRCh38
rs776674677 Chr.20: 56450705-56450705 on Build GRCh38
rs1004096116 Chr.20: 56450661-56450661 on Build GRCh38
rs372643953 Chr.20: 56450707-56450707 on Build GRCh38
rs780985837 Chr.20: 56450761-56450761 on Build GRCh38
rs1031193027 Chr.20: 56450792-56450792 on Build GRCh38
rs376902758 Chr.20: 56450718-56450718 on Build GRCh38
rs765282389 Chr.20: 56450708-56450708 on Build GRCh38
rs200024288 Chr.20: 56451022-56451022 on Build GRCh38
rs953543941 Chr.20: 56450711-56450711 on Build GRCh38
rs777385899 Chr.20: 56450912-56450912 on Build GRCh38
rs373921934 Chr.20: 56451010-56451010 on Build GRCh38
rs113539482 Chr.20: 56451035-56451035 on Build GRCh38
rs974019449 Chr.20: 56450916-56450916 on Build GRCh38
rs987012556 Chr.20: 56450861-56450861 on Build GRCh38
rs543496780 Chr.20: 56451036-56451036 on Build GRCh38
rs563971662 Chr.20: 56451056-56451056 on Build GRCh38
rs745886142 Chr.20: 56450686-56450686 on Build GRCh38
rs1052597145 Chr.20: 56451011-56451011 on Build GRCh38
rs770991801 Chr.20: 56450667-56450667 on Build GRCh38
rs569265583 Chr.20: 56450695-56450695 on Build GRCh38
rs966322313 Chr.20: 56450677-56450677 on Build GRCh38
rs754541747 Chr.20: 56450637-56450637 on Build GRCh38
rs545954619 Chr.20: 56450805-56450805 on Build GRCh38
rs185629257 Chr.20: 56450908-56450908 on Build GRCh38
rs1031009950 Chr.20: 56451066-56451066 on Build GRCh38
rs746052282 Chr.20: 56451021-56451021 on Build GRCh38
rs766064182 Chr.20: 56450627-56450627 on Build GRCh38
rs6069753 Chr.20: 56450706-56450706 on Build GRCh38
rs920580053 Chr.20: 56450976-56450976 on Build GRCh38
rs575043057 Chr.20: 56450750-56450750 on Build GRCh38
rs561677276 Chr.20: 56450938-56450938 on Build GRCh38
rs756466731 Chr.20: 56450659-56450659 on Build GRCh38
rs886589948 Chr.20: 56450585-56450585 on Build GRCh38
rs554430172 Chr.20: 56450772-56450772 on Build GRCh38
rs577479252 Chr.20: 56450800-56450800 on Build GRCh38
rs1036045218 Chr.20: 56450798-56450798 on Build GRCh38
rs750995912 Chr.20: 56450634-56450634 on Build GRCh38
rs903958336 Chr.20: 56451032-56451032 on Build GRCh38
rs570282121 Chr.20: 56450564-56450564 on Build GRCh38
rs796465072 Chr.20: 56451021-56451021 on Build GRCh38
rs545196231 Chr.20: 56450804-56450804 on Build GRCh38
rs1011403828 Chr.20: 56450955-56450955 on Build GRCh38
rs1031397464 Chr.20: 56450630-56450630 on Build GRCh38
rs555287088 Chr.20: 56450741-56450741 on Build GRCh38
rs940414829 Chr.20: 56450767-56450767 on Build GRCh38
rs894339373 Chr.20: 56450949-56450949 on Build GRCh38
rs1056117687 Chr.20: 56450911-56450911 on Build GRCh38
rs148258264 Chr.20: 56450762-56450762 on Build GRCh38
rs750092776 Chr.20: 56450584-56450584 on Build GRCh38
rs527600923 Chr.20: 56451009-56451009 on Build GRCh38
rs200864987 Chr.20: 56451021-56451022 on Build GRCh38
rs967722569 Chr.20: 56450758-56450758 on Build GRCh38
rs774662466 Chr.20: 56450680-56450680 on Build GRCh38
rs553638054 Chr.20: 56450941-56450941 on Build GRCh38
rs562440608 Chr.20: 56450906-56450906 on Build GRCh38
rs1021914003 Chr.20: 56450739-56450739 on Build GRCh38
rs868613416 Chr.20: 56450642-56450642 on Build GRCh38
rs886494096 Chr.20: 56451047-56451047 on Build GRCh38
rs558608299 Chr.20: 56450663-56450663 on Build GRCh38
rs758070857 Chr.20: 56450595-56450595 on Build GRCh38
rs748695905 Chr.20: 56450655-56450655 on Build GRCh38
rs1047869688 Chr.20: 56450575-56450575 on Build GRCh38
rs781529808 Chr.20: 56450651-56450651 on Build GRCh38
rs576027453 Chr.20: 56450907-56450907 on Build GRCh38
rs571413631 Chr.20: 56450680-56450681 on Build GRCh38
rs773078792 Chr.20: 56450717-56450717 on Build GRCh38
rs765902094 Chr.20: 56450723-56450723 on Build GRCh38
rs761676974 Chr.20: 56450566-56450566 on Build GRCh38

View Full Product Details