Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132077710-132078251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs924442672 Chr.X: 132078050-132078050 on Build GRCh38
rs745887910 Chr.X: 132078031-132078031 on Build GRCh38
rs180870479 Chr.X: 132078015-132078015 on Build GRCh38
rs764622962 Chr.X: 132078110-132078110 on Build GRCh38
rs772834154 Chr.X: 132078056-132078056 on Build GRCh38
rs751858239 Chr.X: 132078146-132078146 on Build GRCh38
rs746083026 Chr.X: 132078212-132078212 on Build GRCh38
rs774411143 Chr.X: 132078049-132078049 on Build GRCh38
rs755557174 Chr.X: 132077984-132077984 on Build GRCh38
rs773550469 Chr.X: 132077828-132077828 on Build GRCh38
rs767395087 Chr.X: 132078053-132078053 on Build GRCh38
rs771052393 Chr.X: 132077854-132077854 on Build GRCh38
rs757487294 Chr.X: 132078185-132078185 on Build GRCh38
rs868852708 Chr.X: 132078213-132078213 on Build GRCh38
rs748139262 Chr.X: 132078230-132078230 on Build GRCh38
rs757029488 Chr.X: 132077879-132077879 on Build GRCh38
rs375272641 Chr.X: 132077919-132077919 on Build GRCh38
rs1007286079 Chr.X: 132078120-132078120 on Build GRCh38
rs747281325 Chr.X: 132078018-132078018 on Build GRCh38
rs781449121 Chr.X: 132078201-132078201 on Build GRCh38
rs748600073 Chr.X: 132077994-132077994 on Build GRCh38
rs903939499 Chr.X: 132078203-132078203 on Build GRCh38
rs776699473 Chr.X: 132078029-132078029 on Build GRCh38
rs1056754349 Chr.X: 132078008-132078008 on Build GRCh38
rs372323281 Chr.X: 132078051-132078051 on Build GRCh38
rs909645486 Chr.X: 132077738-132077738 on Build GRCh38
rs779622270 Chr.X: 132077987-132077987 on Build GRCh38
rs766061063 Chr.X: 132078063-132078063 on Build GRCh38
rs1039105692 Chr.X: 132078223-132078223 on Build GRCh38
rs375348707 Chr.X: 132078152-132078152 on Build GRCh38
rs185757626 Chr.X: 132078027-132078027 on Build GRCh38
rs928267747 Chr.X: 132077905-132077905 on Build GRCh38
rs892719233 Chr.X: 132078084-132078084 on Build GRCh38
rs763954895 Chr.X: 132077860-132077860 on Build GRCh38
rs748187461 Chr.X: 132078213-132078218 on Build GRCh38
rs376187248 Chr.X: 132077859-132077859 on Build GRCh38
rs745846839 Chr.X: 132077948-132077950 on Build GRCh38
rs371318658 Chr.X: 132077829-132077829 on Build GRCh38
rs769848562 Chr.X: 132078041-132078041 on Build GRCh38
rs780344859 Chr.X: 132077884-132077884 on Build GRCh38
rs779971174 Chr.X: 132078219-132078219 on Build GRCh38
rs779024567 Chr.X: 132078140-132078141 on Build GRCh38
rs933190542 Chr.X: 132078083-132078083 on Build GRCh38
rs749522497 Chr.X: 132078044-132078044 on Build GRCh38
rs768719723 Chr.X: 132077958-132077959 on Build GRCh38
rs758786935 Chr.X: 132078116-132078116 on Build GRCh38
rs751994722 Chr.X: 132078218-132078218 on Build GRCh38
rs201913554 Chr.X: 132078103-132078103 on Build GRCh38
rs367938621 Chr.X: 132077869-132077869 on Build GRCh38
rs753528994 Chr.X: 132078085-132078085 on Build GRCh38
rs753297140 Chr.X: 132078112-132078112 on Build GRCh38
rs754604228 Chr.X: 132078093-132078093 on Build GRCh38
rs1041565174 Chr.X: 132078030-132078030 on Build GRCh38
rs766008987 Chr.X: 132077939-132077939 on Build GRCh38
rs369513628 Chr.X: 132077957-132077957 on Build GRCh38
rs1025804857 Chr.X: 132077785-132077785 on Build GRCh38
rs1025726284 Chr.X: 132078225-132078225 on Build GRCh38
rs778234656 Chr.X: 132078138-132078138 on Build GRCh38
rs767207907 Chr.X: 132077901-132077901 on Build GRCh38
rs143128232 Chr.X: 132078111-132078111 on Build GRCh38
rs947185883 Chr.X: 132077999-132077999 on Build GRCh38
rs372458379 Chr.X: 132077872-132077872 on Build GRCh38
rs758837654 Chr.X: 132077998-132077998 on Build GRCh38
rs746502563 Chr.X: 132077942-132077950 on Build GRCh38
rs780046824 Chr.X: 132077779-132077779 on Build GRCh38
rs749754497 Chr.X: 132077807-132077810 on Build GRCh38
rs776646478 Chr.X: 132077858-132077858 on Build GRCh38
rs755805433 Chr.X: 132077924-132077924 on Build GRCh38
rs1043882590 Chr.X: 132078168-132078168 on Build GRCh38
rs760433969 Chr.X: 132078058-132078058 on Build GRCh38

View Full Product Details