Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46671891-46672428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS35
Entrez Gene ID
55737
Gene Name
VPS35 retromer complex component
Gene Aliases
MEM3, PARK17
UniGene
Hs.454528
Gene Chromosome Location
Chr.16: 46657979-46689232 on Build GRCh38
Transcripts
NM_018206.5 XM_005256045.3 XM_011523227.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970764 c.1291G>A p.E431K Chr.16: 46672342-46672342 on Build GRCh38
4797553 c.1325G>A p.S442N Chr.16: 46672308-46672308 on Build GRCh38
6397824 c.1297A>G p.R433G Chr.16: 46672336-46672336 on Build GRCh38
5614353 c.1231A>G p.N411D Chr.16: 46672402-46672402 on Build GRCh38
4833507 c.1272C>T p.L424L Chr.16: 46672361-46672361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573966103 Chr.16: 46672168-46672168 on Build GRCh38
rs774333363 Chr.16: 46672223-46672223 on Build GRCh38
rs762332686 Chr.16: 46672369-46672369 on Build GRCh38
rs775375862 Chr.16: 46672235-46672235 on Build GRCh38
rs919945313 Chr.16: 46671990-46671990 on Build GRCh38
rs1026974904 Chr.16: 46672329-46672329 on Build GRCh38
rs377066996 Chr.16: 46672252-46672252 on Build GRCh38
rs368876504 Chr.16: 46672343-46672343 on Build GRCh38
rs958827980 Chr.16: 46671961-46671961 on Build GRCh38
rs753137936 Chr.16: 46672280-46672280 on Build GRCh38
rs878896207 Chr.16: 46672184-46672184 on Build GRCh38
rs770980841 Chr.16: 46672220-46672220 on Build GRCh38
rs143688112 Chr.16: 46672342-46672342 on Build GRCh38
rs908703549 Chr.16: 46672017-46672017 on Build GRCh38
rs952923240 Chr.16: 46671991-46671991 on Build GRCh38
rs920978170 Chr.16: 46672107-46672107 on Build GRCh38
rs187425753 Chr.16: 46672316-46672316 on Build GRCh38
rs530665086 Chr.16: 46672349-46672349 on Build GRCh38
rs370055356 Chr.16: 46672378-46672378 on Build GRCh38
rs991462243 Chr.16: 46671978-46671978 on Build GRCh38
rs756404549 Chr.16: 46672289-46672289 on Build GRCh38
rs1041219131 Chr.16: 46672092-46672092 on Build GRCh38
rs191165923 Chr.16: 46672336-46672336 on Build GRCh38
rs185968939 Chr.16: 46672187-46672187 on Build GRCh38
rs373075116 Chr.16: 46672365-46672365 on Build GRCh38
rs372967908 Chr.16: 46672250-46672250 on Build GRCh38
rs767864451 Chr.16: 46672271-46672271 on Build GRCh38
rs932415832 Chr.16: 46672113-46672113 on Build GRCh38
rs143866394 Chr.16: 46672004-46672004 on Build GRCh38
rs183091274 Chr.16: 46672248-46672248 on Build GRCh38
rs745804093 Chr.16: 46672230-46672230 on Build GRCh38
rs116572885 Chr.16: 46671925-46671925 on Build GRCh38
rs1013031689 Chr.16: 46672323-46672323 on Build GRCh38
rs764924463 Chr.16: 46672356-46672356 on Build GRCh38
rs771840136 Chr.16: 46672234-46672234 on Build GRCh38
rs1036146817 Chr.16: 46671946-46671946 on Build GRCh38
rs190607633 Chr.16: 46672249-46672249 on Build GRCh38
rs371991621 Chr.16: 46672256-46672256 on Build GRCh38
rs375087873 Chr.16: 46672371-46672371 on Build GRCh38
rs893494821 Chr.16: 46672179-46672179 on Build GRCh38
rs908749023 Chr.16: 46672344-46672344 on Build GRCh38
rs1042729762 Chr.16: 46672208-46672208 on Build GRCh38
rs772703701 Chr.16: 46671968-46671968 on Build GRCh38
rs555625377 Chr.16: 46672028-46672028 on Build GRCh38
rs376209552 Chr.16: 46672306-46672306 on Build GRCh38
rs745893720 Chr.16: 46672320-46672320 on Build GRCh38
rs576005754 Chr.16: 46672262-46672262 on Build GRCh38
rs139200616 Chr.16: 46672341-46672341 on Build GRCh38
rs749503515 Chr.16: 46672300-46672300 on Build GRCh38
rs944244357 Chr.16: 46672034-46672034 on Build GRCh38
rs17850640 Chr.16: 46672275-46672275 on Build GRCh38
rs778105954 Chr.16: 46672293-46672293 on Build GRCh38
rs760344479 Chr.16: 46672247-46672247 on Build GRCh38
rs757800220 Chr.16: 46672229-46672230 on Build GRCh38

View Full Product Details