Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 224445-224590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHA
Entrez Gene ID
6389
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Gene Aliases
CMD1GG, FP, PGL5, SDH1, SDH2, SDHF
UniGene
Hs.440475
Gene Chromosome Location
Chr.5: 218223-264816 on Build GRCh38
Transcripts
NM_001294332.1 NM_004168.3 XM_005248331.3 XM_011514072.1 XM_011514073.1 XM_017009685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994269 c.262G>A p.A88T Chr.5: 224471-224471 on Build GRCh38
3787051 c.264A>G p.A88A Chr.5: 224473-224473 on Build GRCh38
4854337 c.251G>A p.G84E Chr.5: 224460-224460 on Build GRCh38
6384337 c.245A>T p.E82V Chr.5: 224454-224454 on Build GRCh38
3761191 c.309A>G p.A103A Chr.5: 224518-224518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs786205145 Chr.5: 224504-224504 on Build GRCh38
rs771169259 Chr.5: 224537-224537 on Build GRCh38
rs763941248 Chr.5: 224532-224532 on Build GRCh38
rs745395045 Chr.5: 224506-224506 on Build GRCh38
rs761532792 Chr.5: 224534-224534 on Build GRCh38
rs575715550 Chr.5: 224568-224568 on Build GRCh38
rs373601972 Chr.5: 224566-224566 on Build GRCh38
rs746449707 Chr.5: 224571-224571 on Build GRCh38
rs200519133 Chr.5: 224465-224465 on Build GRCh38
rs138016874 Chr.5: 224475-224475 on Build GRCh38
rs377620054 Chr.5: 224496-224496 on Build GRCh38
rs774504151 Chr.5: 224540-224540 on Build GRCh38
rs771662494 Chr.5: 224509-224509 on Build GRCh38
rs371061737 Chr.5: 224555-224555 on Build GRCh38
rs1139424 Chr.5: 224518-224518 on Build GRCh38
rs767673132 Chr.5: 224545-224545 on Build GRCh38
rs371274523 Chr.5: 224499-224499 on Build GRCh38
rs876658637 Chr.5: 224520-224521 on Build GRCh38
rs375298039 Chr.5: 224485-224485 on Build GRCh38
rs545120389 Chr.5: 224570-224570 on Build GRCh38
rs776529046 Chr.5: 224536-224536 on Build GRCh38
rs778267118 Chr.5: 224484-224484 on Build GRCh38
rs764449603 Chr.5: 224556-224556 on Build GRCh38
rs201953257 Chr.5: 224569-224569 on Build GRCh38
rs756543943 Chr.5: 224469-224469 on Build GRCh38
rs747036101 Chr.5: 224525-224525 on Build GRCh38
rs886060514 Chr.5: 224478-224478 on Build GRCh38
rs370899204 Chr.5: 224533-224533 on Build GRCh38
rs765674382 Chr.5: 224567-224567 on Build GRCh38
rs754258872 Chr.5: 224562-224562 on Build GRCh38
rs111533078 Chr.5: 224541-224541 on Build GRCh38
rs752714156 Chr.5: 224552-224552 on Build GRCh38
rs768467351 Chr.5: 224529-224529 on Build GRCh38

View Full Product Details