Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMG7
Entrez Gene ID
9887
Gene Name
SMG7, nonsense mediated mRNA decay factor
Gene Aliases
C1orf16, EST1C, SGA56M
Chromosome Location
Chr.1: 183471962 - 183554193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174061.1 NP_001167532.1 8 - 9 1218 73
NM_173156.2 NP_775179.1 9 - 10 1250 73
NM_201568.2 NP_963862.1 9 - 10 1250 73
NM_201569.2 NP_963863.2 9 - 10 1250 73
XM_005245648.3 XP_005245705.1 11 - 12 1935 73
XM_005245649.3 XP_005245706.1 10 - 11 1798 73
XM_005245652.3 XP_005245709.1 9 - 10 1119 73
XM_005245653.4 XP_005245710.1 10 - 11 1221 73
XM_006711676.3 XP_006711739.1 10 - 11 1798 73
XM_006711678.3 XP_006711741.1 9 - 10 1119 73
XM_011510205.2 XP_011508507.1 12 - 13 2113 73
XM_011510206.2 XP_011508508.1 12 - 13 2113 73
XM_011510207.2 XP_011508509.1 10 - 11 1837 73
XM_011510208.2 XP_011508510.1 12 - 13 2113 73
XM_011510209.2 XP_011508511.1 12 - 13 2113 73
XM_011510210.2 XP_011508512.1 8 - 9 1089 73
XM_017002966.1 XP_016858455.1 11 - 12 1390 73
XM_017002967.1 XP_016858456.1 11 - 12 1398 73
XM_017002968.1 XP_016858457.1 8 - 9 1093 73
XM_017002969.1 XP_016858458.1 9 - 10 1312 73
XM_017002970.1 XP_016858459.1 9 - 10 1236 73
XM_017002971.1 XP_016858460.1 9 - 10 1312 73
GenBank mRNA AB085674.1 - 9 - 10 1063 73
AK056035.1 - 9 - 10 1119 73
AK074297.1 - 9 - 10 1107 73
AK091477.1 - 8 - 9 1087 73
AK297207.1 - 10 - 11 1282 73
AK299178.1 - 8 - 9 1089 73
AK307394.1 - 9 - 10 1111 73
AY386363.1 - 2 - 3 219 73
BC036381.1 - 9 - 10 1118 73
BC052565.1 - 2 - 3 245 73
D87437.2 - 9 - 10 1094 73

View Full Product Details