The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174061.1 | NP_001167532.1 | 12 - 13 | 1636 | 144 |
NM_173156.2 | NP_775179.1 | 13 - 14 | 1668 | 144 | |
NM_201568.2 | NP_963862.1 | 13 - 14 | 1668 | 144 | |
NM_201569.2 | NP_963863.2 | 13 - 14 | 1668 | 144 | |
XM_005245648.3 | XP_005245705.1 | 15 - 16 | 2353 | 144 | |
XM_005245649.3 | XP_005245706.1 | 14 - 15 | 2216 | 144 | |
XM_005245652.3 | XP_005245709.1 | 13 - 14 | 1537 | 144 | |
XM_005245653.4 | XP_005245710.1 | 14 - 15 | 1639 | 144 | |
XM_006711676.3 | XP_006711739.1 | 14 - 15 | 2216 | 144 | |
XM_006711678.3 | XP_006711741.1 | 13 - 14 | 1537 | 144 | |
XM_011510205.2 | XP_011508507.1 | 16 - 17 | 2531 | 144 | |
XM_011510206.2 | XP_011508508.1 | 16 - 17 | 2531 | 144 | |
XM_011510207.2 | XP_011508509.1 | 14 - 15 | 2255 | 144 | |
XM_011510208.2 | XP_011508510.1 | 16 - 17 | 2531 | 144 | |
XM_011510209.2 | XP_011508511.1 | 16 - 17 | 2531 | 144 | |
XM_011510210.2 | XP_011508512.1 | 12 - 13 | 1507 | 144 | |
XM_017002966.1 | XP_016858455.1 | 15 - 16 | 1808 | 144 | |
XM_017002967.1 | XP_016858456.1 | 15 - 16 | 1816 | 144 | |
XM_017002968.1 | XP_016858457.1 | 12 - 13 | 1511 | 144 | |
XM_017002969.1 | XP_016858458.1 | 13 - 14 | 1730 | 144 | |
XM_017002970.1 | XP_016858459.1 | 13 - 14 | 1654 | 144 | |
XM_017002971.1 | XP_016858460.1 | 13 - 14 | 1730 | 144 | |
GenBank mRNA | AB085674.1 | - | 13 - 14 | 1481 | 144 |
AK056035.1 | - | 13 - 14 | 1537 | 144 | |
AK074297.1 | - | 13 - 14 | 1525 | 144 | |
AK091477.1 | - | 12 - 13 | 1505 | 144 | |
AK297207.1 | - | 14 - 15 | 1700 | 144 | |
AK299178.1 | - | 12 - 13 | 1507 | 144 | |
AK307394.1 | - | 13 - 14 | 1529 | 144 | |
AL833675.1 | - | 1 - 2 | 51 | 144 | |
AY386363.1 | - | 6 - 7 | 637 | 144 | |
BC036381.1 | - | 13 - 14 | 1536 | 144 | |
BC052565.1 | - | 6 - 7 | 663 | 144 | |
D87437.2 | - | 13 - 14 | 1512 | 144 |