The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308096.1 | NP_001295025.1 | 11 - 12 | 1038 | 94 |
NM_001324096.1 | NP_001311025.1 | 10 - 11 | 881 | 94 | |
NM_001324097.1 | NP_001311026.1 | 10 - 11 | 899 | 94 | |
NM_001324098.1 | NP_001311027.1 | 10 - 11 | 1091 | 94 | |
NM_032569.3 | NP_115958.2 | 11 - 12 | 1056 | 94 | |
NR_136695.1 | 12 - 13 | 1341 | 94 | ||
NR_136696.1 | 11 - 12 | 1124 | 94 | ||
NR_136697.1 | 11 - 12 | 1142 | 94 | ||
NR_136698.1 | 10 - 11 | 1073 | 94 | ||
NR_136699.1 | 10 - 11 | 1091 | 94 | ||
XM_005255639.4 | XP_005255696.2 | 9 - 10 | 806 | 94 | |
XM_011522716.2 | XP_011521018.1 | 11 - 12 | 1184 | 94 | |
XM_011522717.1 | XP_011521019.1 | 12 - 13 | 1241 | 94 | |
XM_017023787.1 | XP_016879276.1 | 10 - 11 | 1068 | 94 | |
XM_017023788.1 | XP_016879277.1 | 12 - 13 | 1223 | 94 | |
XM_017023789.1 | XP_016879278.1 | 11 - 12 | 1190 | 94 | |
GenBank mRNA | AF244907.1 | - | 11 - 12 | 1037 | 94 |
AF326966.1 | - | 11 - 12 | 1001 | 94 | |
AK296842.1 | - | 10 - 11 | 790 | 94 | |
AK307834.1 | - | 11 - 12 | 1031 | 94 | |
AY352585.1 | - | 11 - 12 | 1019 | 94 | |
BC003693.1 | - | 9 - 10 | 930 | 94 | |
BC032855.1 | - | 11 - 12 | 1047 | 94 | |
BC047223.1 | - | 11 - 12 | 1036 | 94 | |
BC064940.1 | - | 12 - 13 | 1240 | 94 |