Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf40
Entrez Gene ID
283461
Gene Name
chromosome 12 open reading frame 40
Gene Aliases
HEL-206, HEL-S-94
Chromosome Location
Chr.12: 39626167 - 39908300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031748.3 NP_001026918.2 11 - 12 1583 142
NR_135051.1 11 - 12 1636 142
XM_005268806.3 XP_005268863.1 10 - 11 1393 142
XM_011538231.2 XP_011536533.1 10 - 11 1620 142
GenBank mRNA AK097445.1 - 11 - 12 1585 142
BC048120.1 - 11 - 12 1567 142
EU668334.1 - 11 - 12 1585 142
GQ472216.1 - 11 - 12 1585 142

View Full Product Details