Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46038477-46038750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1 XM_017024489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4891361 c.2510G>A p.S837N Chr.17: 46038569-46038569 on Build GRCh38
1743592 c.2470C>T p.R824* Chr.17: 46038609-46038609 on Build GRCh38
334060 c.2410G>T p.D804Y Chr.17: 46038669-46038669 on Build GRCh38
7001119 c.2466_2491del26 p.V823fs*8 Chr.17: 46038588-46038613 on Build GRCh38
6580515 c.2495C>G p.S832C Chr.17: 46038584-46038584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141409854 Chr.17: 46038514-46038514 on Build GRCh38
rs773238363 Chr.17: 46038599-46038599 on Build GRCh38
rs143653891 Chr.17: 46038598-46038598 on Build GRCh38
rs759562366 Chr.17: 46038702-46038702 on Build GRCh38
rs148718200 Chr.17: 46038541-46038541 on Build GRCh38
rs200903355 Chr.17: 46038589-46038589 on Build GRCh38
rs1004017370 Chr.17: 46038695-46038695 on Build GRCh38
rs375461096 Chr.17: 46038499-46038499 on Build GRCh38
rs780829206 Chr.17: 46038642-46038642 on Build GRCh38
rs777630407 Chr.17: 46038630-46038630 on Build GRCh38
rs769338654 Chr.17: 46038578-46038578 on Build GRCh38
rs754964384 Chr.17: 46038539-46038539 on Build GRCh38
rs758243969 Chr.17: 46038555-46038555 on Build GRCh38
rs372105810 Chr.17: 46038656-46038656 on Build GRCh38
rs796052597 Chr.17: 46038545-46038545 on Build GRCh38
rs776867963 Chr.17: 46038700-46038700 on Build GRCh38
rs1037817208 Chr.17: 46038574-46038574 on Build GRCh38
rs535801827 Chr.17: 46038683-46038683 on Build GRCh38
rs776349500 Chr.17: 46038570-46038570 on Build GRCh38
rs539976549 Chr.17: 46038662-46038662 on Build GRCh38
rs752702501 Chr.17: 46038618-46038618 on Build GRCh38
rs751168223 Chr.17: 46038634-46038634 on Build GRCh38
rs762076648 Chr.17: 46038516-46038516 on Build GRCh38
rs746462091 Chr.17: 46038506-46038506 on Build GRCh38
rs935626079 Chr.17: 46038573-46038573 on Build GRCh38
rs201526313 Chr.17: 46038665-46038665 on Build GRCh38
rs766447665 Chr.17: 46038527-46038527 on Build GRCh38
rs753966599 Chr.17: 46038601-46038601 on Build GRCh38
rs765044321 Chr.17: 46038616-46038616 on Build GRCh38
rs751450813 Chr.17: 46038716-46038716 on Build GRCh38
rs200086055 Chr.17: 46038629-46038629 on Build GRCh38
rs772747352 Chr.17: 46038505-46038505 on Build GRCh38
rs768530197 Chr.17: 46038512-46038515 on Build GRCh38
rs200079722 Chr.17: 46038538-46038538 on Build GRCh38
rs746835116 Chr.17: 46038719-46038719 on Build GRCh38
rs754877424 Chr.17: 46038615-46038615 on Build GRCh38
rs184281762 Chr.17: 46038666-46038666 on Build GRCh38
rs770500282 Chr.17: 46038569-46038569 on Build GRCh38
rs773681267 Chr.17: 46038673-46038673 on Build GRCh38
rs765371986 Chr.17: 46038703-46038703 on Build GRCh38
rs757031050 Chr.17: 46038638-46038638 on Build GRCh38
rs758071565 Chr.17: 46038624-46038624 on Build GRCh38
rs554142966 Chr.17: 46038709-46038709 on Build GRCh38
rs774638418 Chr.17: 46038515-46038515 on Build GRCh38
rs775683727 Chr.17: 46038710-46038710 on Build GRCh38
rs12150090 Chr.17: 46038520-46038520 on Build GRCh38
rs746688275 Chr.17: 46038566-46038566 on Build GRCh38
rs771181281 Chr.17: 46038692-46038692 on Build GRCh38
rs777242394 Chr.17: 46038559-46038559 on Build GRCh38
rs767677684 Chr.17: 46038519-46038519 on Build GRCh38
rs927360678 Chr.17: 46038575-46038575 on Build GRCh38
rs747933631 Chr.17: 46038550-46038550 on Build GRCh38
rs749802464 Chr.17: 46038577-46038577 on Build GRCh38
rs774750158 Chr.17: 46038584-46038584 on Build GRCh38
rs11539538 Chr.17: 46038600-46038600 on Build GRCh38
rs770587918 Chr.17: 46038507-46038507 on Build GRCh38
rs763785963 Chr.17: 46038715-46038715 on Build GRCh38
rs368839053 Chr.17: 46038513-46038513 on Build GRCh38
rs570967215 Chr.17: 46038655-46038655 on Build GRCh38

View Full Product Details