The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184963.1 | NP_001171892.1 | 1 - 2 | 50 | 54 |
NM_173160.2 | NP_775183.1 | 1 - 2 | 50 | 54 | |
GenBank mRNA | AY358584.1 | - | 1 - 2 | 50 | 54 |
BC054876.1 | - | 1 - 2 | 56 | 54 | |
BG432244.1 | - | 1 - 2 | 53 | 54 |