The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174937.3 | NP_777597.2 | 7 - 8 | 1281 | 78 |
XM_011539577.1 | XP_011537879.1 | 8 - 9 | 1314 | 78 | |
GenBank mRNA | AK096269.1 | - | 7 - 8 | 3058 | 78 |
BC042951.1 | - | 7 - 8 | 1286 | 78 | |
BC093639.1 | - | 7 - 8 | 1130 | 78 | |
BC101535.1 | - | 7 - 8 | 1130 | 78 |