The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174975.4 | NP_777635.1 | 1 - 2 | 139 | 58 |
XM_011530128.2 | XP_011528430.1 | 1 - 2 | 107 | 58 | |
GenBank mRNA | AY158086.1 | - | 1 - 2 | 50 | 58 |
AY240872.1 | - | 1 - 2 | 139 | 58 | |
BC069641.1 | - | 1 - 2 | 62 | 58 | |
BC101004.1 | - | 1 - 2 | 78 | 58 | |
BU584962.1 | - | 1 - 2 | 60 | 58 |