Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA9
Entrez Gene ID
327657
Gene Name
serpin family A member 9
Gene Aliases
GCET1, SERPINA11, SERPINA11b
Chromosome Location
Chr.14: 94462721 - 94476333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042518.1 NP_001035983.1 1 - 2 196 85
NM_175739.3 NP_783866.2 1 - 2 196 85
GenBank mRNA AK301082.1 - 1 - 2 90 85
AY185496.1 - 1 - 2 196 85
AY220118.1 - 1 - 2 110 85
AY220119.1 - 1 - 2 74 85
AY358700.1 - 1 - 2 40 85
BC111498.1 - 1 - 2 99 85

View Full Product Details