The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042518.1 | NP_001035983.1 | 3 - 4 | 546 | 95 |
NM_001284275.1 | NP_001271204.1 | 2 - 3 | 656 | 95 | |
NM_001284276.1 | NP_001271205.1 | 3 - 4 | 553 | 95 | |
NM_175739.3 | NP_783866.2 | 2 - 3 | 846 | 95 | |
XM_011536714.2 | XP_011535016.1 | 2 - 3 | 952 | 95 | |
XM_011536715.2 | XP_011535017.1 | 3 - 4 | 816 | 95 | |
XM_011536716.2 | XP_011535018.1 | 2 - 3 | 762 | 95 | |
XM_011536717.1 | XP_011535019.1 | 2 - 3 | 1645 | 95 | |
GenBank mRNA | AK301082.1 | - | 2 - 3 | 95 | |
AY185496.1 | - | 2 - 3 | 846 | 95 | |
AY185497.1 | - | 2 - 3 | 656 | 95 | |
AY220121.1 | - | 3 - 4 | 95 | ||
AY358700.1 | - | 2 - 3 | 690 | 95 | |
BC111498.1 | - | 2 - 3 | 749 | 95 |