The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319053.1 | NP_001305982.1 | 20 - 21 | 2660 | 60 |
NM_001470.3 | NP_001461.1 | 20 - 21 | 2724 | 60 | |
NM_021903.2 | NP_068703.1 | 15 - 16 | 2524 | 60 | |
NM_021904.3 | NP_068704.2 | 19 - 20 | 2538 | 60 | |
XM_005248982.2 | XP_005249039.1 | 20 - 21 | 2539 | 60 | |
XM_006715047.3 | XP_006715110.1 | 19 - 20 | 2339 | 60 | |
XM_011514453.2 | XP_011512755.1 | 18 - 19 | 2318 | 60 | |
XM_017010676.1 | XP_016866165.1 | 16 - 17 | 2070 | 60 | |
GenBank mRNA | AB208790.1 | - | 20 - 21 | 2459 | 60 |
AF099148.1 | - | 20 - 21 | 2546 | 60 | |
AJ012185.1 | - | 19 - 20 | 2438 | 60 | |
AJ012186.1 | - | 15 - 16 | 2087 | 60 | |
AJ012187.1 | - | 18 - 19 | 2252 | 60 | |
AJ012288.1 | - | 20 - 21 | 2481 | 60 | |
AJ225028.1 | - | 20 - 21 | 2672 | 60 | |
AJ225029.1 | - | 15 - 16 | 2266 | 60 | |
AK223619.1 | - | 15 - 16 | 2309 | 60 | |
AK290191.1 | - | 15 - 16 | 2309 | 60 | |
AK312308.1 | - | 15 - 16 | 2305 | 60 | |
BC041332.2 | - | 15 - 16 | 2309 | 60 | |
BC042598.1 | - | 15 - 16 | 2298 | 60 | |
BC050532.1 | - | 15 - 16 | 2296 | 60 | |
Y11044.1 | - | 19 - 20 | 2435 | 60 |