The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001470.3 | NP_001461.1 | 4 - 5 | 758 | 87 |
XM_005248982.2 | XP_005249039.1 | 4 - 5 | 573 | 87 | |
XM_011514455.1 | XP_011512757.1 | 4 - 5 | 573 | 87 | |
GenBank mRNA | AF099148.1 | - | 4 - 5 | 580 | 87 |
AF301005.1 | - | 3 - 4 | 475 | 87 | |
AJ012185.1 | - | 3 - 4 | 472 | 87 | |
AJ012288.1 | - | 4 - 5 | 515 | 87 | |
AJ225028.1 | - | 4 - 5 | 706 | 87 | |
Y11044.1 | - | 3 - 4 | 469 | 87 |