The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001812.2 | NP_001803.2 | 11 - 12 | 2238 | 110 |
XM_006714064.2 | XP_006714127.1 | 11 - 12 | 2301 | 110 | |
XM_011531542.2 | XP_011529844.1 | 11 - 12 | 2301 | 110 | |
GenBank mRNA | AK307928.1 | - | 11 - 12 | 2203 | 110 |
AK308170.1 | - | 11 - 12 | 2203 | 110 | |
AK308214.1 | - | 2 - 3 | 360 | 110 | |
BC030695.1 | - | 12 - 13 | 2542 | 110 | |
BC041117.1 | - | 11 - 12 | 2198 | 110 | |
BX648166.1 | - | 11 - 12 | 2113 | 110 | |
M95724.1 | - | 11 - 12 | 2211 | 110 |