Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20650351-20650618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PINK1
Entrez Gene ID
65018
Gene Name
PTEN induced putative kinase 1
Gene Aliases
BRPK, PARK6
UniGene
Hs.389171
Gene Chromosome Location
Chr.1: 20633455-20651511 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Gene Symbol
PINK1-AS
Entrez Gene ID
100861548
Gene Name
PINK1 antisense RNA
Gene Aliases
NAPINK1, PINK1-AS1, PINK1AS
UniGene
Hs.710259
Gene Chromosome Location
Chr.1: 20642657-20652193 on Build GRCh38
Transcripts
NM_032409.2 NR_046507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6451148 c.1519C>T p.L507F Chr.1: 20650464-20650464 on Build GRCh38
1929013 c.1609G>A p.A537T Chr.1: 20650554-20650554 on Build GRCh38
5422934 c.1640A>C p.K547T Chr.1: 20650585-20650585 on Build GRCh38
425200 c.1615T>G p.L539V Chr.1: 20650560-20650560 on Build GRCh38
903283 c.1638G>T p.E546D Chr.1: 20650583-20650583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764454206 Chr.1: 20650505-20650505 on Build GRCh38
rs765320312 Chr.1: 20650502-20650502 on Build GRCh38
rs761156393 Chr.1: 20650582-20650582 on Build GRCh38
rs753034026 Chr.1: 20650424-20650424 on Build GRCh38
rs746598031 Chr.1: 20650451-20650451 on Build GRCh38
rs886045848 Chr.1: 20650523-20650523 on Build GRCh38
rs774261081 Chr.1: 20650480-20650480 on Build GRCh38
rs562390614 Chr.1: 20650382-20650382 on Build GRCh38
rs730880302 Chr.1: 20650518-20650519 on Build GRCh38
rs753932833 Chr.1: 20650403-20650404 on Build GRCh38
rs566377750 Chr.1: 20650577-20650577 on Build GRCh38
rs531477772 Chr.1: 20650518-20650518 on Build GRCh38
rs757382757 Chr.1: 20650437-20650438 on Build GRCh38
rs760962342 Chr.1: 20650499-20650499 on Build GRCh38
rs536023801 Chr.1: 20650570-20650570 on Build GRCh38
rs768447159 Chr.1: 20650454-20650454 on Build GRCh38
rs1031736135 Chr.1: 20650413-20650413 on Build GRCh38
rs771032673 Chr.1: 20650554-20650554 on Build GRCh38
rs755432820 Chr.1: 20650407-20650407 on Build GRCh38
rs554043854 Chr.1: 20650576-20650576 on Build GRCh38
rs779418622 Chr.1: 20650449-20650449 on Build GRCh38
rs779294259 Chr.1: 20650455-20650455 on Build GRCh38
rs780954418 Chr.1: 20650538-20650538 on Build GRCh38
rs773504394 Chr.1: 20650459-20650459 on Build GRCh38
rs749131400 Chr.1: 20650553-20650553 on Build GRCh38
rs1013932116 Chr.1: 20650487-20650487 on Build GRCh38
rs1043424 Chr.1: 20650507-20650507 on Build GRCh38
rs869165879 Chr.1: 20650500-20650500 on Build GRCh38
rs371223801 Chr.1: 20650390-20650390 on Build GRCh38
rs778495491 Chr.1: 20650432-20650432 on Build GRCh38
rs61744200 Chr.1: 20650447-20650447 on Build GRCh38
rs764654160 Chr.1: 20650585-20650585 on Build GRCh38
rs529830769 Chr.1: 20650431-20650431 on Build GRCh38
rs763284341 Chr.1: 20650461-20650461 on Build GRCh38
rs149466252 Chr.1: 20650494-20650494 on Build GRCh38
rs938160611 Chr.1: 20650574-20650574 on Build GRCh38
rs367949045 Chr.1: 20650475-20650475 on Build GRCh38
rs149070918 Chr.1: 20650550-20650550 on Build GRCh38
rs146126901 Chr.1: 20650549-20650549 on Build GRCh38
rs779798626 Chr.1: 20650524-20650524 on Build GRCh38
rs368454614 Chr.1: 20650391-20650391 on Build GRCh38
rs758615369 Chr.1: 20650582-20650583 on Build GRCh38
rs371338035 Chr.1: 20650521-20650521 on Build GRCh38
rs768105319 Chr.1: 20650421-20650421 on Build GRCh38
rs141363209 Chr.1: 20650489-20650489 on Build GRCh38
rs751715929 Chr.1: 20650398-20650398 on Build GRCh38
rs757843763 Chr.1: 20650448-20650448 on Build GRCh38
rs750664040 Chr.1: 20650541-20650542 on Build GRCh38
rs748103970 Chr.1: 20650546-20650546 on Build GRCh38
rs560377198 Chr.1: 20650456-20650456 on Build GRCh38
rs1026284739 Chr.1: 20650374-20650374 on Build GRCh38
rs527565484 Chr.1: 20650457-20650457 on Build GRCh38
rs61749520 Chr.1: 20650566-20650566 on Build GRCh38
rs371881185 Chr.1: 20650406-20650406 on Build GRCh38
rs751012180 Chr.1: 20650525-20650525 on Build GRCh38
rs754543494 Chr.1: 20650528-20650528 on Build GRCh38

View Full Product Details