The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004065.2 | NP_004056.2 | 1 - 1 | 1087 | 86 |
GenBank mRNA | BC113472.1 | - | 1 - 1 | 1012 | 86 |
BC113474.1 | - | 1 - 1 | 1012 | 86 | |
M16965.1 | - | 1 - 1 | 953 | 86 |