The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244959.1 | NP_001231888.1 | 3 - 4 | 500 | 86 |
NM_001244960.1 | NP_001231889.1 | 3 - 4 | 276 | 86 | |
NM_174938.5 | NP_777598.3 | 3 - 4 | 500 | 86 | |
XM_017014588.1 | XP_016870077.1 | 3 - 4 | 190 | 86 | |
GenBank mRNA | AK094281.1 | - | 3 - 4 | 201 | 86 |
AK223597.1 | - | 3 - 4 | 427 | 86 | |
AK297722.1 | - | 3 - 4 | 418 | 86 | |
AY137774.1 | - | 3 - 4 | 284 | 86 | |
EF560742.1 | - | 3 - 4 | 351 | 86 |